
4-Methoxy-3-propoxybenzoic acid
Katalogové číslo APOSOR0589-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:210.23 g/mol
Melting Pt:150…153 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:5922-57-6
Formula:C₁₁H₁₄O₄
MDL Number:MFCD00831063