
2-Iodobenzenesulphonyl chloride
Catalog # APOSOR0548-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:302.52 g/mol
Boiling Pt:135 °C/1mmHg
Melting Pt:48…51 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
CAS Number:63059-29-0
Formula:C₆H₄ClIO₂S
MDL Number:MFCD00833336