
2-(1,4-Diazepan-1-yl)ethanol
Catalog # APOSOR0518-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:144.22 g/mol
Boiling Pt:116…120 °C/1mmHg
Storage Temperature:Ambient
UN:3267
ADR:8,III
:GHS05
CAS Number:53427-65-9
Formula:C₇H₁₆N₂O
MDL Number:MFCD01565842