

2,6-Dichloro-3-nitropyridine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:16013-85-7
:
Formula:C₅H₂Cl₂N₂O₂
MDL Number:MFCD00006234
:192.99 g/mol
Melting Pt:55…60 °C
Flash Pt:110 °C
EINECS:240-151-1
:GHS07
:GHS08
Storage Temperature:Ambient
Frequently Bought Together