

Ethyl-4-bromobenzoate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5798-75-4
:
Formula:C₉H₉BrO₂
MDL Number:MFCD00016329
:229.07 g/mol
Boiling Pt:262…264 °C
Density:1.403 g/cm³ (25 °C)
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:227-347-2
UN:3082
ADR:9+EHS,III
:GHS07
:GHS09
Frequently Bought Together