
4'-(1-Piperazinyl)acetophenone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:51639-48-6
:
Formula:C₁₂H₁₆N₂O
MDL Number:MFCD00005962
:204.27 g/mol
Melting Pt:107…110 °C
EINECS:257-332-6
:GHS07
Storage Temperature:Ambient
Frequently Bought Together