Order Entry
Czech Republic
ContactUsLinkComponent
4-Hexylphenylhydrazine hydrochloride 95%
Irritant
4-Hexylphenylhydrazine hydrochloride 95%
Catalog # APOSOR0201-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Hexylphenylhydrazine hydrochloride 95%
Catalog # APOSOR0201-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
Melting Pt:219…221 °C (dec.)
 :GHS07
Storage Temperature:Ambient
CAS Number:126062-51-9
Formula:C₁₂H₂₁ClN₂
MDL Number:MFCD06245514