

5-Oxo-5,6,7,8-tetrahydronaphthalene-2-carboxylic acid 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3470-46-0
:
Formula:C₁₁H₁₀O₃
MDL Number:MFCD01763813
:190.2 g/mol
Melting Pt:215…222 °C (dec.)
UN:1759
ADR:8,III
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together