Order Entry
Czech Republic
ContactUsLinkComponent
2-(4-Methylsulphonylphenyl)ethylamine hydrochloride 95+%
Irritant
2-(4-Methylsulphonylphenyl)ethylamine hydrochloride 95+%
Catalog # APOSOR0178-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(4-Methylsulphonylphenyl)ethylamine hydrochloride 95+%
Catalog # APOSOR0178-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :235.73 g/mol
Melting Pt:202…205 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:849020-96-8
Formula:C₉H₁₄ClNO₂S
MDL Number:MFCD06245499