1-(2,4-Dichlorophenyl)ethylamine 98%
Catalog # APOSOR01762-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:190.07 g/mol
Storage Temperature:Ambient
CAS Number:89981-75-9
Formula:C₈H₉Cl₂N
MDL Number:MFCD05215236