Order Entry
Czech Republic
ContactUsLinkComponent
2-Methyl-1H-benzoimidazole-5-carboxylic acid 98%
Irritant
2-Methyl-1H-benzoimidazole-5-carboxylic acid 98%
Catalog # APOSOR01727-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Methyl-1H-benzoimidazole-5-carboxylic acid 98%
Catalog # APOSOR01727-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :176.18 g/mol
Melting Pt:300…314 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:709-19-3
Formula:C₉H₈N₂O₂
MDL Number:MFCD00800488