
8-Methyl-8-azabicyclo[3.2.1]octan-3-amine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:98998-25-5
:
Formula:C₈H₁₆N₂
MDL Number:MFCD00210699
:140.23 g/mol
Boiling Pt:42…45 °C/0.75mmHg
:GHS07
Storage Temperature:Ambient
Frequently Bought Together