
1-(4-Biphenylyl)ethylamine
Catalog # APOSOR01696-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:197.28 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:86217-82-5
Formula:C₁₄H₁₅N
MDL Number:MFCD02667819