Order Entry
Czech Republic
ContactUsLinkComponent
2-(4-Methoxyphenyl)piperazine
Corrosive
Irritant
2-(4-Methoxyphenyl)piperazine
Catalog # APOSOR01693-50MG
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
2-(4-Methoxyphenyl)piperazine
Catalog # APOSOR01693-50MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :192.26 g/mol
Storage Temperature:Ambient
UN:3259
ADR:8,III
 :GHS05
 :GHS07
CAS Number:91517-26-9
Formula:C₁₁H₁₆N₂O
MDL Number:MFCD05864677