Order Entry
Czech Republic
ContactUsLinkComponent
4'-Bromo-2'-chloroacetanilide
Irritant
4'-Bromo-2'-chloroacetanilide
Catalog # APOSOR015905-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4'-Bromo-2'-chloroacetanilide
Catalog # APOSOR015905-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Melting Pt:158…160 °C
 :248.51 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:3460-23-9
Formula:H₃CC(O)NH(C₆H₃BrCl)
MDL Number:MFCD00040852