
5-Nitro-o-cresol 98%
Catalog # APOSOR0108-500G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:153.14 g/mol
Boiling Pt:180 °C/15mmHg
Melting Pt:111…115 °C
Storage Temperature:Ambient
EINECS:226-580-7
UN:2446
ADR:6.1,III
:GHS06
CAS Number:5428-54-6
Formula:C₇H₇NO₃
MDL Number:MFCD00043909