Order Entry
Czech Republic
ContactUsLinkComponent
p-Toluenesulphinic acid sodium salt
Irritant
p-Toluenesulphinic acid sodium salt
Catalog # APOSOR0042-500G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
p-Toluenesulphinic acid sodium salt
Catalog # APOSOR0042-500G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :178.19 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:824-79-3
 :MFCD00149640
Formula:C₇H₇NaO₂S
MDL Number:MFCD00013136