Order Entry
Czech Republic
ContactUsLinkComponent
2,4-Dibromobenzenesulphonyl chloride 98%
Corrosive
2,4-Dibromobenzenesulphonyl chloride 98%
Catalog # APOSOR0035-5G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
2,4-Dibromobenzenesulphonyl chloride 98%
Catalog # APOSOR0035-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :334.42 g/mol
Boiling Pt:128 °C/0.2mmHg
Melting Pt:82…86 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
 :GHS05
CAS Number:72256-95-2
Formula:C₆H₃Br₂ClO₂S
MDL Number:MFCD03093759