
4'-Chloro-3'-nitroacetophenone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:5465-65-6
:
Formula:C₈H₆ClNO₃
MDL Number:MFCD00007083
:199.59
Melting Pt:99…101 °C
EINECS:226-769-4
:GHS07
Storage Temperature:Ambient
Frequently Bought Together