
4-Butylbenzenesulphonyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:54997-92-1
:
Formula:C₁₀H₁₃ClO₂S
MDL Number:MFCD00173908
:232.73 g/mol
Boiling Pt:126…128 °C/0.6mmHg
Melting Pt:13…15 °C
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05
Frequently Bought Together