Order Entry
Czech Republic
ContactUsLinkComponent
Yttrium (III) nitrate hexahydrate 99,9%
Oxidizing
Irritant
Yttrium (III) nitrate hexahydrate 99,9%
Catalog # APOSIN3856-100G
Supplier:  Apollo Scientific
CAS Number:  
Oxidizing
Irritant
Yttrium (III) nitrate hexahydrate 99,9%
Catalog # APOSIN3856-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :383.01 g/mol
Density:2.682 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:233-802-6
UN:1477
ADR:5.1,II
 :GHS03
 :GHS07
CAS Number:13494-98-9
Formula:N₃O₉Y·6H₂O
MDL Number:MFCD00149940