Order Entry
Czech Republic
ContactUsLinkComponent
Ytterbium triacetate tetrahydrate 99,9%
Ytterbium triacetate tetrahydrate 99,9%
Catalog # APOSIN3787-5G
Supplier:  Apollo Scientific
CAS Number:  
Ytterbium triacetate tetrahydrate 99,9%
Catalog # APOSIN3787-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :422.23 g/mol
Density:2.09 g/cm³ (25 °C)
Storage Temperature:Ambient
CAS Number:15280-58-7
Formula:C₆H₉O₆Yb·4H₂O