Titanium(IV) oxide rutile 99.999%
Catalog # APOSIN3709-50G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:79.88 g/mol
Melting Pt:1855 °C
Density:4.17 g/cm³ (25 °C)
Storage Temperature:Ambient
CAS Number:1317-80-2
Formula:O₂Ti
MDL Number:MFCD00011269