Samarium trinitrate hexahydrate 99,9%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:13759-83-6
:24581-35-9
Formula:N₃O₉Sm·6H₂O
MDL Number:MFCD00149857
:444.47 g/mol
Melting Pt:78 °C
Density:2.375 g/cm³ (25 °C)
Storage Temperature:Ambient
Frequently Bought Together