
Neodymium trichloride hexahydrate 99,9%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:13477-89-9
:
Formula:Cl₃Nd·6H₂O
MDL Number:MFCD00149801
:358.69 g/mol
Melting Pt:124 °C
Density:2.282 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient
Frequently Bought Together