
Iron(II) bromide, anhydrous
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7789-46-0
:
Formula:Br₂Fe
MDL Number:MFCD00016081
Boiling Pt:934 °C
Melting Pt:684 °C
Density:4,636 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:232-168-8
:GHS07
Frequently Bought Together