Order Entry
Czech Republic
ContactUsLinkComponent
Gallium trinitrate hydrate 99,9%
Gallium trinitrate hydrate 99,9%
Catalog # APOSIN1885-25G
Supplier:  Apollo Scientific
CAS Number:  
Gallium trinitrate hydrate 99,9%
Catalog # APOSIN1885-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :273.75 g/mol
Melting Pt:110 °C
Storage Temperature:Ambient
EINECS:236-815-5
UN:1477
ADR:5.1,II
 :GHS07
CAS Number:69365-72-6
Formula:GaN₃O₉·H₂O
MDL Number:MFCD00149723