


Copper(II) bromide, Reagent Grade
Catalog # APOSIN1540-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:223.35 g/mol
Boiling Pt:900 °C
Melting Pt:498 °C
Density:4.71 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:232-167-2
UN:3260
ADR:8,III
:GHS05
:GHS07
:GHS09
CAS Number:7789-45-9
Formula:Br₂Cu
MDL Number:MFCD00010970