Order Entry
Czech Republic
ContactUsLinkComponent
Cerium trinitrate hexahydrate 99,9%
Oxidizing
Irritant
Cerium trinitrate hexahydrate 99,9%
Catalog # APOSIN1423-100G
Supplier:  Apollo Scientific
CAS Number:  
Oxidizing
Irritant
Cerium trinitrate hexahydrate 99,9%
Catalog # APOSIN1423-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :434.22 g/mol
Boiling Pt:200 °C
Storage Temperature:Ambient
EINECS:233-297-2
UN:1477
ADR:5.1,III
 :GHS03
 :GHS07
CAS Number:10294-41-4
Formula:Ce(NO₃)₃·6H₂O
MDL Number:MFCD00149631