Order Entry
Czech Republic
ContactUsLinkComponent
Barium thiocyanate trihydrate 99%
Barium thiocyanate trihydrate 99%
Katalogové číslo APOSIN1225-25G
Supplier:  Apollo Scientific
CAS Number:  
Barium thiocyanate trihydrate 99%
Katalogové číslo APOSIN1225-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Density:2.286 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:1564
ADR:6.1,III
 :307.55 g/mol (253.49 anhy)
CAS Number:68016-36-4
Formula:Ba(SCN)₂·3H₂O
MDL Number:MFCD00150182