
D-(+)-Sucrose
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:57-50-1
:
Formula:C₁₂H₂₂O₁₁
MDL Number:MFCD00006626
Melting Pt:185…190 °C
Density:1.58 g/cm³ (25 °C)
:342.3 g/mol
EINECS:200-334-9
:GHS07
Storage Temperature:Ambient
Frequently Bought Together