
Sulpho-SANPAH (Sulphosuccinimidyl-6-(4′-azido-2′-nitrophenylamino)hexanoate)
Catalog # APOSBIPA109-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:492.40 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:102568-43-4
Formula:C₁₆H₁₇O₉N₆NaS