Order Entry
Czech Republic
ContactUsLinkComponent
 
 
POPSO (β,β'-dihydroxypiperazine-1,4-dipropanesulphonic acid) ≥99%
  APOSBIP0705-25G
 :  
 
POPSO (β,β'-dihydroxypiperazine-1,4-dipropanesulphonic acid) ≥99%
  APOSBIP0705-25G
 :  

 

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

 :
 :300 °C
 :362.4203 g/mol
 :GHS07
 :Ambient
 :68189-43-5
 :C₁₀H₂₂N₂O₈S₂
 :MFCD00038350