Nile red
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 9-(Diethylamino)-5H-benzo[a]phenoxazin-5-one
- CAS Number:7385-67-3
- Formula:C₂₀H₁₈N₂O₂
- MDL Number:MFCD00011639
- Molecular Weight:318.38 g/mol
- Melting Pt:203…205 °C
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
Synonyms: 9-(Diethylamino)-5H-benzo[a]phenoxazin-5-one
- CAS Number:7385-67-3
- Formula:C₂₀H₁₈N₂O₂
- MDL Number:MFCD00011639
- Molecular Weight:318.38 g/mol
- Melting Pt:203…205 °C
- Storage Temperature:Ambient
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



