
4-Nitrophenyl-β-D-glucuronide ≥99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:10344-94-2
:
Formula:C₁₂H₁₃NO₉
MDL Number:MFCD00036210
:315.23 g/mol
EINECS:233-753-0
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together