Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
4-Methylumbelliferyl alpha-D-mannopyranoside
Catalog # APOSBIM1100-25MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Methylumbelliferyl alpha-D-mannopyranoside
Catalog # APOSBIM1100-25MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :338.31 g/mol
EINECS:249-073-2
 :GHS07
Storage Temperature:Freezer
CAS Number:28541-83-5
Formula:C₁₆H₁₈O₈
MDL Number:MFCD00067662