
4-Methylumbelliferyl-alpha-D-glucopyranoside
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:17833-43-1
:
Formula:C₁₆H₁₈O₈
MDL Number:MFCD00067661
:338.32 g/mol
EINECS:241-794-0
:GHS07
Storage Temperature:Freezer
Frequently Bought Together