Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Iminothiolane hydrochloride (Traut's Reagent)
2-Iminothiolane hydrochloride (Traut's Reagent)
2-Iminothiolane hydrochloride (Traut's Reagent)
2-Iminothiolane hydrochloride (Traut's Reagent)
APOSBIM099-1G
:
Apollo Scientific
:
2-Iminothiolane hydrochloride (Traut's Reagent)
APOSBIM099-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
137.625 g/mol
:
GHS07
:
Refrigerator
:
4781-83-3
:
C₄H₈ClNS
:
MFCD00039013