
L(+)-Rhamnose monohydrate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:10030-85-0
:
Formula:C₆H₁₂O₅·1H₂O
MDL Number:MFCD00149363
:182.17 g/mol
Melting Pt:90 °C
EINECS:222-793-4
:GHS07
Storage Temperature:Ambient
Frequently Bought Together