HEPPSO (β-hydroxy-4-(2-hydroxyethyl)piperazine-1-propanesulphonic acid) ≥98%
Catalog # APOSBIH0893-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Typ balení:Bottle
- Bal.:100 g
- Sekvence:3401
Specifications
About this item
:
:268.3305 g/mol
Storage Temperature:Ambient
CAS Number:68399-78-0
Formula:C₉H₂₀N₂O₅S
MDL Number:MFCD00038351