D-(+)-Galactosamine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1772-03-8
:
Formula:C₆H₁₄ClNO₅
MDL Number:MFCD00135830
:215.64 g/mol
Melting Pt:172…180 °C
Storage Temperature:Refrigerator
EINECS:217-198-1
Frequently Bought Together