

N-(3-Ethynylphenyl)-6,7-bis(2-methoxyethoxy)quinazolin-4-amine hydrochloride ≥99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:183319-69-9
:
Formula:C₂₂H₂₄ClN₃O₄
MDL Number:MFCD07781272
:429.901 g/mol
Melting Pt:215…227 °C
:GHS07
:GHS09
Storage Temperature:Freezer
Frequently Bought Together