
2-Deoxy-D-galactose ≥99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1949-89-9
:
Formula:C₆H₁₂O₅
MDL Number:MFCD00014649
:164.16 g/mol
Melting Pt:107…110 °C
EINECS:217-765-3
:GHS07
Storage Temperature:Ambient
Frequently Bought Together