6A-Azido-6A-deoxy-beta-cyclodextrin
Bottle, 1 g
Catalog # APOSBICY1079-1G
Supplier: Apollo Scientific
6A-Azido-6A-deoxy-beta-cyclodextrin
Catalog # APOSBICY1079-1G
Supplier: Apollo Scientific
CAS Number: 98169-85-8
Jump to:
Product Details & Documents
Synonyms: Mono(6-azido-6-desoxy)-β-cyclodextrin, Mono-6A-azido-β-cyclodextrin, 6-Deoxy-6-azido-β-cyclodextrin, 6-Monoazido-6-monodeoxy-β-cyclodextrin, 6-Azido-6-deoxycyclomaltoheptaose, 6-Monoazido-6-deoxy-β-cyclodextrin, 6-Monodeoxy-6-monoazido-beta-cyclodextrin, 2,4,7,9,12,14,17,19,22,24,27,29,32,34-Tetradecaoxaoctacyclo[31.2.2.23,6.28,11.213,16.218,21.223,26.228,31]nonatetracontane, β-cyclodextrin derivative, 6-Monoazido-β-cyclodextrin, 6A-Azido-6A-deoxy-β-cyclodextrin
- CAS Number:98169-85-8
- Formula:C₄₂H₆₉N₃O₃₄
- MDL Number:MFCD05864973
- Molecular Weight:1160.00 g/mol
- Melting Pt:206 °C (dec.)
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:1 g



