Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Domů
Heptakis-6-iodo-6-deoxy-beta-cyclodextrin
Heptakis-6-iodo-6-deoxy-β-cyclodextrin
Heptakis-6-iodo-6-deoxy-β-cyclodextrin
Heptakis-6-iodo-6-deoxy-β-cyclodextrin
APOSBICY1019-5G
:
Apollo Scientific
:
Heptakis-6-iodo-6-deoxy-β-cyclodextrin
APOSBICY1019-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
1904.28 g/mol
:
Ambient
:
30754-23-5
:
C₄₂H₆₃I₇O₂₈
:
MFCD00803290