Mono-6-O-(p-toluenesulphonyl)-β-cyclodextrin
Catalog # APOSBICY1015-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:1289.1706 g/mol
Storage Temperature:Ambient
CAS Number:67217-55-4
Formula:C₄₉H₇₆O₃₇S
MDL Number:MFCD05864975