1,2-O-Isopropylidene-ɑ-D-glucurono-6,3-lactone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:20513-98-8
:MFCD01863407
Formula:C₉H₁₂O₆
MDL Number:MFCD00061641
:216.19 g/mol
Storage Temperature:Ambient
Frequently Bought Together