
(+)-1-Deoxynojirimycin hydrochloride 98% min
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:73465-43-7
:
Formula:C₆H₁₄ClNO₄
MDL Number:MFCD00083611
:199.64 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together