
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:36051-68-0
:
Formula:C₉H₁₄N₃Na₂O₁₄P₃
MDL Number:MFCD00078193
:527.12 g/mol
Flash Pt:113 °C
:GHS07
Storage Temperature:Freezer
Frequently Bought Together