cis-4-Azido-N-(tert-butoxycarbonyl)-L-proline ≥98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:132622-65-2
:
Formula:C₁₀H₁₆N₄O₄
MDL Number:MFCD04115791
:256.26 g/mol
Storage Temperature:Ambient
Frequently Bought Together